missing translation for 'onlineSavingsMsg'
Learn More

REEP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 14643-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16883384

  • 112.71 € / 20 Mikroliter

Verfügbar ab
17-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

REEP5 (receptor expression-enhancing protein 5), also known as C5orf18, DP1, TB2 or D5S346, is a 189 amino acid multi-pass membrane protein. Thought to promote the functional cell surface expression of olfactory receptors, REEP5 belongs to the DP1 family and is encoded by a gene that maps to chromosome 5. With 181 million base pairs encoding around 1,000 genes, chromosome 5 is about 6% of human genomic DNA. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the parm of chromosome 5 leads to cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
Spezifikation

Spezifikation

REEP5
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B2RZ37, Q00765, Q60870
C5orf18, D5S346, DP1, Polyposis locus protein 1, Protein TB2, receptor accessory protein 5, REEP5, TB2
Rabbit
IgG
20 μL
-20°C
Primary
13476, 364838, 7905
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Flow Cytometry
Unconjugated
REEP5
Liquid
Reep5
REEP5 Fusion Protein Ag6229
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation