missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Spezifikation
Antigen | REEP5 |
---|---|
Konzentration | 0.3 mg/mL |
Anwendungen | Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Flow Cytometry |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16883384
|
Proteintech
14643-1-AP-20UL |
20 μL |
140.00 €
20 Mikroliter |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16873384
|
Proteintech
14643-1-AP-150UL |
150 μL |
399.00 €
150 Mikroliter |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
REEP5 (receptor expression-enhancing protein 5), also known as C5orf18, DP1, TB2 or D5S346, is a 189 amino acid multi-pass membrane protein. Thought to promote the functional cell surface expression of olfactory receptors, REEP5 belongs to the DP1 family and is encoded by a gene that maps to chromosome 5. With 181 million base pairs encoding around 1,000 genes, chromosome 5 is about 6% of human genomic DNA. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the parm of chromosome 5 leads to cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.Spezifikation
REEP5 | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
B2RZ37, Q00765, Q60870 | |
13476, 364838, 7905 | |
REEP5 Fusion Protein Ag6229 | |
Primary | |
-20°C | |
REEP5 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C5orf18, D5S346, DP1, Polyposis locus protein 1, Protein TB2, receptor accessory protein 5, REEP5, TB2 | |
Reep5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |