Beschreibung
RelB Monoclonal antibody specifically detects RelB in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry
Spezifikation
Spezifikation
| Antigen | RelB |
| Anwendungen | Flow Cytometry, ELISA, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Klon | 9E4 |
| Konjugat | Unconjugated |
| Verdünnung | Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Zusammensetzung | PBS, pH 7.4, 150mM NaCl and 50% glycerol |
| Gen-Alias | I-Rel, IREL, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, v-rel reticuloendotheliosis viral oncogene homolog B |
| Wirtsspezies | Rabbit |
| Immunogen | A synthesized peptide derived from Human RelB [UniProt Q01201] |
| Mehr anzeigen |