missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RELM alpha Monoclonal Antibody (DS8RELM), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 46-5441-80
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Rat Monoclonal Antibody
RELM alpha Monoclonal antibody specifically detects RELM alpha in Mouse samples. It is validated for Flow Cytometry.Spezifikation
RELM alpha | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9EP95 | |
Retnla | |
Recombinant mouse RELM alpha protein | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
DS8RELM | |
PerCP-eFluor 710 | |
Retnla | |
1810019L16Rik; cysteine-rich secreted protein A12-gamma; Cysteine-rich secreted protein FIZZ1; Fizz1; Fizz-1; FIZZ2; found in inflammatory zone 1; Himf; HXCP2; Hypoxia-induced mitogenic factor; parasite-induced macrophage novel gene 1 protein; Pmng1; RELMa; RELMalpha; RELM-alpha; RELMb; RELMbeta; RELM-beta; resistin like alpha; resistin-like alpha; Retnla; Xcp2 | |
Rat | |
Affinity chromatography | |
RUO | |
57262 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |