missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
REPS1 Polyclonal specifically detects REPS1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | REPS1 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.5 mg/ml |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1.0 ug/ml |
| Zusammensetzung | PBS, 2% Sucrose |
| Gen-Alias | RALBP1, RALBP1 associated Eps domain containing 1, ralBP1-associated Eps domain-containing protein 1, RalBP1-interacting protein 1 |
| Wirtsspezies | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human REPS1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?