Learn More
Restek™ Frits for Agilent™ HPLC Systems (Models 1050, 1100, 1200)
Product Code. 10073514
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent HPLC Systems (Models 1100, 1050, 1200)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10073514 Agilent HPLC Systems (Models 1100, 1050, 1200) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10073514 Supplier Restek™ Supplier No. 25466
Verfügbar ab: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Frits for Agilent™ HPLC Systems (Models 1050, 1100, 1200)
  • Meets or exceeds OEM performance
Specifications
For Use With (Equipment) Agilent HPLC Systems (Models 1100, 1050, 1200)
Material PTFE
Type Frit