missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Siltek™ Press-Tight™ Connectors

Siltek deactivation for an inert pathway that maintains sample integrity

Marke:  Restek™ 20449

Weitere Details : CAS : 000-00-0 Netto-Gewicht : 1.00000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13021790

  • 479.88 € / 25 Stück


Verfügbar ab
19-11-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Made from inert fused silica
  • For use in connecting guard columns to analytical columns
  • Fit Restek column outer diameters from 0.33 to 0.74mm (0.1 to 0.53mm inside diameter)

  • Angled Press-Tight connector approximates the curvature of a capillary column to reduce strain on column-end connections

Spezifikation

Spezifikation

25/Pk.
Siltek
Universal Press-Tight Connector
Videos
Dokumentation