Learn More
Rhodium(III) Chloride Hydrate, TRC
Product Code. 30418976
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418976 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30418976 Supplier Toronto Research Chemicals Supplier No. TRCR318645500MG
 Restricted Item
Verfügbar ab: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Rhodium(III) Chloride Hydrate
CAS 20765-98-4
Molecular Formula Cl3Rh.xH2O
Quantity 500 mg
Synonym Rhodium Trichloride Hydrate
InChI Formula InChI=1S/C21H27F3O8S/c1-5-6-7-10-15(25)11-8-9-14(2)31-20(26)19-17(30-4)12-16(29-3)13-18(19)32-33(27,28)21(22,23)24/h5,12-14H,1,6-11H2,2-4H3
SMILES COc1cc(OC)c(C(=O)OC(C)CCCC(=O)CCCC=C)c(OS(=O)(=O)C(F)(F)F)c1
IUPAC Name 6-oxoundec-10-en-2-yl 2,4-dimethoxy-6-(trifluoromethylsulfonyloxy)benzoate
Molecular Weight (g/mol) 209.26
Formula Weight 496.1379