missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RIMS3 Polyclonal antibody specifically detects RIMS3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | RIMS3 |
| Anwendungen | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | DyLight 594 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | KIAA0137, KIAA0237, Nim3, NIM3Rab-3 interacting molecule 3, Rab-3-interacting molecule 3, regulating synaptic membrane exocytosis 3, regulating synaptic membrane exocytosis protein 3, RIM 3, RIM3, RIM3 gamma |
| Wirtsspezies | Rabbit |
| Immunogen | Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human Rim3. The immunogen is located within amino acids 60 - 110 of Rim3. Amino Acid Squence: LPQPEGATKKLRSNIRR |
| Reinigungsverfahren | Peptide affinity purified |
| Menge | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?