missing translation for 'onlineSavingsMsg'
Learn More

RIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 16388-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16885994

  • 112.71 € / 20 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Rit and its neuron-specific homologue, Rin, define a recently discovered subfamily of Ras-related GTPases. Rit and Rin are membrane-associated in spite of the fact that they lack a CAAX box or similar C-terminal lipidation motif. Rit and Rin display 64% amino acid sequence identity and share a unique nine amino acid effector domain (DPTIEDAYK) that is 100% conserved between the murine and human proteins. Although the effector domain sequences of Rit and Rin are very similar to that of Ras, Rit and Rin have been shown to interact with the known Ras-binding proteins RalGDS, Rlf and AF-6, but not the Raf kinases, RIN1 or the p110 subunit of PI3 kinase. For this reason, it has been suggested that Rit and Rin may play important roles in the regulation of signaling pathways distinct from those controlled by Ras.
Spezifikation

Spezifikation

RIN1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97680, Q13671, Q921Q7
Ras and Rab interactor 1, Ras inhibitor JC99, RIN1
Rabbit
IgG
20 μL
-20°C
Primary
207119, 225870, 9610
Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RIN1
Liquid
RIN1
RIN1 Fusion Protein Ag9451
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation