missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RLIM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16121-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
RNF12 is a RING-H2 zinc finger protein. It has been shown to be a ubiquitin protein ligase that targets LIM domain binding 1 (LDB1/CLIM), and causes proteasome-dependent degradation of LDB1. This protein and LDB1 are co-repressors of LHX1/LIM-1, a homeodomain transcription factor. Alternatively spliced transcript variants encoding the same protein have been reported.Spezifikation
RLIM | |
Polyclonal | |
Unconjugated | |
RLIM | |
NY REN 43, R LIM, RING finger protein 12, RLIM, RNF12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108348168, 19820, 51132 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NVW2, Q9WTV7 | |
RLIM | |
RLIM Fusion Protein Ag9077 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |