missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Spezifikation
Antigen | RNASE3 |
---|---|
Konzentration | 0.32 mg/mL |
Anwendungen | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16896815
|
Proteintech
55338-1-AP-20UL |
20 μL |
140.00 €
20 Mikroliter |
Verfügbar ab: 24-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16886815
|
Proteintech
55338-1-AP-150UL |
150 μL |
399.00 €
150 Mikroliter |
Verfügbar ab: 24-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
ECP (eosinophil cationic protein), also known as RNS3 or RNASE3 (ribonuclease,RNase A family, 3), is a 160 amino acid potent cytotoxic secretory protein that belongs to the pancreatic ribonuclease family. ECP localizes to the granule matrix of the eosinophil and is involved in the immune response system. ECP is a cytotoxin, neurotoxin and helminthotoxin that is secreted by activated human eosinophils. ECP exhibits anti-parasitic, antibacterial and ribonucleolytic activities. It has been suggested that ECP induces the neurotoxic effect known as the Gordon phenomenon, a syndrome manifested by ataxia, muscular rigidity, paralysis and tremor that may lead to death. ECP is considered a marker of eosinophilic inflammation and high levels have been found in cases of active asthma and other allergic diseases.Spezifikation
RNASE3 | |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
O35290, P12724 | |
53876, 6037 | |
Peptide | |
Primary | |
-20°C | |
RNASE3 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ECP, Eosinophil cationic protein, Ribonuclease 3, RNase 3, RNASE3, RNS3 | |
Ear3, RNASE3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |