missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55338-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ECP (eosinophil cationic protein), also known as RNS3 or RNASE3 (ribonuclease,RNase A family, 3), is a 160 amino acid potent cytotoxic secretory protein that belongs to the pancreatic ribonuclease family. ECP localizes to the granule matrix of the eosinophil and is involved in the immune response system. ECP is a cytotoxin, neurotoxin and helminthotoxin that is secreted by activated human eosinophils. ECP exhibits anti-parasitic, antibacterial and ribonucleolytic activities. It has been suggested that ECP induces the neurotoxic effect known as the Gordon phenomenon, a syndrome manifested by ataxia, muscular rigidity, paralysis and tremor that may lead to death. ECP is considered a marker of eosinophilic inflammation and high levels have been found in cases of active asthma and other allergic diseases.Spezifikation
RNASE3 | |
Polyclonal | |
Unconjugated | |
RNASE3 | |
ECP, Eosinophil cationic protein, Ribonuclease 3, RNase 3, RNASE3, RNS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53876, 6037 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35290, P12724 | |
Ear3, RNASE3 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |