missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF144A Polyclonal Antibody

Artikelnummer. 16608054
Change view
Click to view available options
Menge:
100 μL
Unit Size:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16608054 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16608054 Supplier Invitrogen™ Supplier No. PA5101573

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RNF144A.

UBCE7IP4 (Ubiquitin-conjugating enzyme 7- interacting protein 4), also known as RNF144A (RING finger protein 144A), KIAA0161 or RNF144, is a 292 amino acid single-pass membrane protein that contains one RING-type zinc finger and two IBR-type zinc fingers. Functioning as an E3 ubiquitin-protein ligase, UBCE7IP4 accepts ubiquitin (in the form of a thioester) from E2 ubiquitin-conjugating enzymes, such as UBC8 and UBCH7, and transfers that ubiquitin residue to target substrates. Via its RING finger, UBCE7IP4 may play a role in protein-DNA and protein-protein interactions throughout the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF144A
Anwendungen Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen RNF144A
Gen-Zugriffsnummer P50876, Q925F3
Gen-Alias E3 ubiquitin-protein ligase RNF144A; Kiaa0161; probable E3 ubiquitin-protein ligase RNF144A; RGD1561737; ring finger protein 144; ring finger protein 144A; RNF144; RNF144A; Ubce7ip4; UbcM4-interacting protein 4; ubiquitin conjugating enzyme 7 interacting protein 4; ubiquitin-conjugating enzyme 7-interacting protein 4; Uip4
Gensymbole RNF144A
Wirtsspezies Rabbit
Immunogen A synthesized peptide derived from human RNF144A(Accession P50876), corresponding to amino acid residues H151-M201.
Reinigungsverfahren Affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 108089, 9781
Zielspezies Human, Mouse
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.