missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16031-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a molecular chaperone that is a member of the chaperonin containing TCP1 complex , also known as the TCP1 ring complex. This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized. In addition, three pseudogenes that appear to be derived from this gene have been found.Spezifikation
RNF2 | |
Polyclonal | |
Unconjugated | |
RNF2 | |
BAP 1, BAP1, DING, HIP2 interacting protein 3, HIPI3, Protein DinG, RING finger protein 1B, ring finger protein 2, RING finger protein BAP 1, RING1B, RING2, RNF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19821, 304850, 6045 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4KLY4, Q99496, Q9CQJ4 | |
RNF2 | |
RNF2 Fusion Protein Ag8900 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |