missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROCK2, GST Tag Recombinant Protein
ROCK2 Recombinant Human Protein
Marke: Invitrogen PV3759
Weitere Details : Netto-Gewicht : 0.05250kg
Description
ROCK2, the predominant ROCK isoform in skeletal muscle, is an important regulator downstream of Rho GTPases.Specifications
NP_004841 | |
367, 9475 | |
10 μg | |
DMPK Kinase Family | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Ser⁄Thr Kinases: ACG Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
ROCK2 | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
90.8 kDa | |
ROCK2 | |
aa 1-552 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
10 μg | |
Dry Ice | |
N-Terminal | |
Rho-associated coiled-coil containing protein kinase 2 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |