Learn More
RockPhos Pd G3, 98%, BLD Pharm™
Product Code. 31068306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31068306 100 mg
31066419 5 g
31057352 1 g
31056537 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31068306 Supplier BLD Pharm Supplier No. BD00824728100mg
 Restricted Item
Verfügbar ab: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 2009020-38-4
Molecular Formula C44H62NO4PPdS
MDL Number MFCD27952542
Quantity 100 mg
SMILES P(C1C(=CC=C(C=1C1C(=CC(=CC=1C(C)C)C(C)C)C(C)C)C)OC)([Pd+2]1(NC2=CC=CC=C2C2=CC=CC=[C-]12)[O-]S(=O)(=O)C)(C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 838.43
Percent Purity 98%
Chemical Name or Material RockPhos Pd G3