missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROS1 Recombinant Human Protein
ROS1 is a tyrosine protein kinase.
Marke: Invitrogen™ PV4120
Weitere Details : Netto-Gewicht : 0.00900kg
Description
ROS1 functions as a growth or differentiation factor receptor.
Specifications
NP_002935 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
ROS1, ROS, MCF4 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
79.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
SEV Kinase Family | |
Recombinant | |
Baculovirus |