Learn More
BUCHI Rotavapor Vacuum Seal
Product Code. 16275112 Shop All Buchi Products
Change view
Click to view available options
For Use With (Equipment):
R-300, R-210, R-215, R II
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16275112 R-300, R-210, R-215, R II Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16275112 Supplier BUCHI Supplier No. 11069167
Lager vorrätig
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Rotavapor R-300 Vacuum Seal with O-Ring.
  • PTFE Vacuum Seal VS-26
  • Replacement for WD-26
TRUSTED_SUSTAINABILITY
Specifications
Description VS-26
Material PTFE
Type Replacement Seal
For Use With (Equipment) R-300, R-210, R-215, R II