Learn More
Tocris Bioscience™ Rp-8-Br-PET-cGMPS
Beschreibung
Competitive, reversible cGMP-dependent protein kinase (PKG) inhibitor; cGMP analog.
This product is sold in units of 560μg, equivalent to 1μmol.
Spezifikation
Spezifikation
| Chemischer Name oder Material | Rp-8-Br-PET-cGMPS |
| CAS | 185246-32-6 |
| Menge | 560 μg |
| Summenformel | C18H15BrN5NaO6PS |
| Formelmasse | 562.27 |
| Reinheit | >99% |
| Molekulargewicht | 563.275 |