missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL39 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14990-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein L39 (RPL39), also known as 60S Ribosomal Protein L39, consists of 51 amino acids and is the smallest protein found in liver ribosomes. Ribosomal Protein L39 localizes to cytoplasm and is a member of the S39E family of ribosomal proteins. The gene encoding Ribosomal Protein L39 maps to human chromosome Xq24, and shares 92 percent homology with Ribosomal Protein L39L (Ribosomal Protein L39-like), which is also known as RPL39L, L39-2 or RPL39L1. Ribosomal Protein L39L is associated with the 60S ribosomal subunit and is expressed in testis with localization in nucleolus.Spezifikation
RPL39 | |
Polyclonal | |
Unconjugated | |
RPL39 | |
60S ribosomal protein L39, ribosomal protein L39, RPL39 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25347, 6170, 67248 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62891, P62892, P62893 | |
RPL39 | |
RPL39 Fusion Protein Ag6975 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |