missing translation for 'onlineSavingsMsg'
Learn More

RPL8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 16981-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886784

  • 329.85 € / 150 Mikroliter

Verfügbar ab
17-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L2P family of ribosomal proteins. It is located in the cytoplasm. In rat, the protein associates with the 5.8S rRNA, very likely participates in the binding of aminoacyl-tRNA, and is a constituent of the elongation factor 2-binding site at the ribosomal subunit interface. Alternatively spliced transcript variants encoding the same protein exist. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Spezifikation

Spezifikation

RPL8
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62917, P62918, P62919
60S ribosomal protein L8, ribosomal protein L8, RPL8
Rabbit
IgG
150 μL
-20°C
Primary
26961, 26962, 6132
Western Blot
Unconjugated
RPL8
Liquid
RPL8
RPL8 Fusion Protein Ag9957
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation