missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Spezifikation
Antigen | RPS12 |
---|---|
Konzentration | 0.25 mg/mL |
Anwendungen | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16896134
|
Proteintech
16490-1-AP-20UL |
20 μL |
140.00 €
20 Mikroliter |
Verfügbar ab: 23-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16886134
|
Proteintech
16490-1-AP-150UL |
150 μL |
399.00 €
150 Mikroliter |
Verfügbar ab: 23-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
This antibody specifically recognizes the 15 kDa RPS12 protein.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S12E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal cancers compared to matched normal colonic mucosa has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Spezifikation
RPS12 | |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P25398, P63323, P63324 | |
20042, 6206, 65139 | |
RPS12 Fusion Protein Ag9615 | |
Primary | |
-20°C | |
RPS12 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
40S ribosomal protein S12, ribosomal protein S12, RPS12 | |
RPS12 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |