missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RPS6KA4 (MSK2), GST Tag Recombinant Protein
Human RPS6KA4 (MSK2), GST Tag Recombinant Protein
Marke: Invitrogen PV3782
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
RPS6KA4 (MSK2) is a serine⁄threonine protein kinase that is essential for the regulation of various cellular processes. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos.Spezifikation
NP_003933 | |
1812, 8986 | |
10 μg | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: AGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™ | |
ribosomal protein S6 kinase, 90kDa, polypeptide 4 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
113.1 kDa | |
RPS6KA4 (MSK2) | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
RPS6KA4 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |