Beschreibung
RSPH10B Polyclonal antibody specifically detects RSPH10B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | RSPH10B |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Zusammensetzung | PBS (pH 7.2), 40% Glycerol |
| Gen-Zugriffsnummer | B2RC85 |
| Gen-Alias | MGC50833, radial spoke head 10 homolog B, radial spoke head 10 homolog B (Chlamydomonas), radial spoke head 10 homolog B2, RSPH10B2 |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: EGLMHGQGTYIWADGLKYEGDFVKNVPMNHGVYTWPDGSMYEGEVVNGMRNGFGMFKCSTQPVSYIGHWCNGKRHGKGSIYYNQEGTCWYEGDWVQN |
| Mehr anzeigen |