missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RTCD1 Polyclonal antibody specifically detects RTCD1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | RTCD1 |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500-1:2000, ELISA |
| Zusammensetzung | PBS, 50% Glycerol, pH (7.3) |
| Gen-Alias | phosphate cyclase, RNA 3'-terminal phosphate cyclase, RNA cyclase, RNA terminal phosphate cyclase domain 1, RNA terminal phosphate cyclase domain-containing protein 1, RNA-3'-phosphate cyclase, RPC1, RPCEC 6.5.1.4, RTC domain containing 1, RTC domain-containing protein 1, RTC1 |
| Wirtsspezies | Rabbit |
| Immunogen | RNA terminal phosphate cyclase domain 1. (Uniprot# O00442) |
| Reinigungsverfahren | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?