Learn More
Rubitecan, TRC
Product Code. 30412417
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412417 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30412417 Supplier Toronto Research Chemicals Supplier No. TRCR7015002MG
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 9-Nitro-20(S)-Camptothecin
CAS 91421-42-0
Molecular Formula C20 H15 N3 O6
Quantity 2 mg
InChI Formula InChI=1S/C20H15N3O6/c1-2-20(26)13-7-16-17-10(8-22(16)18(24)12(13)9-29-19(20)25)6-11-14(21-17)4-3-5-15(11)23(27)28/h3-7,26H,2,8-9H2,1H3/t20-/m0/s1
SMILES CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(cccc5nc34)[N+](=O)[O-])C2=O
Molecular Weight (g/mol) 393.35
Formula Weight 393.0961