Learn More
Rufinamide, TRC
Product Code. 30003910
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003910 50 mg 50mg
30006526 250 mg 250mg
17903225 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30003910 Supplier Toronto Research Chemicals Supplier No. TRCR70155050MG
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106308-44-5
Molecular Formula C10 H8 F2 N4 O
Molecular Weight (g/mol) 238.19
Synonym 1-[(2,6-Difluorophenyl)methyl]-1H-1,2,3-triazole-4-carboxamide, 1-[(2,6-Difiuorophenyl)methyl]-1H-1,2,3-triazole-4 carboxamide, Banzel, CGP 33101, E 2080, Inovelon, RUF 331, Rufinamide
IUPAC Name 1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
SMILES NC(=O)c1cn(Cc2c(F)cccc2F)nn1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C10H8F2N4O/c11-7-2-1-3-8(12)6(7)4-16-5-9(10(13)17)14-15-16/h1-3,5H,4H2,(H2,13,17)
Formula Weight 238.0666
Percent Purity >95%
Chemical Name or Material Rufinamide
Grade HPLC
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.