missing translation for 'onlineSavingsMsg'
Learn More

Ruzadolane, TRC

Product Code. 30421444
Change view
Click to view available options
Menge:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30421444 500 mg
30412194 250 mg
30415843 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421444 Supplier Toronto Research Chemicals Supplier No. TRCR702990500MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115762-17-9
Summenformel C18H19F2N5S
Molekulargewicht (g/mol) 375.44
Synonym 3-[[2-[4-(2,4-Difluorophenyl)-1-piperazinyl]ethyl]thio]-s-triazolo[4,3-a]pyridine
IUPAC-Name 3-[2-[4-(2,4-difluorophenyl)piperazin-1-yl]ethylsulfanyl]-[1,2,4]triazolo[4,3-a]pyridine
SMILES Fc1ccc(N2CCN(CCSc3nnc4ccccn34)CC2)c(F)c1

Specifications

Chemischer Name oder Material Ruzadolane
Menge 500 mg
InChI-Formel InChI=1S/C18H19F2N5S/c19-14-4-5-16(15(20)13-14)24-9-7-23(8-10-24)11-12-26-18-22-21-17-3-1-2-6-25(17)18/h1-6,13H,7-12H2
Formelmasse 375.1329
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.