missing translation for 'onlineSavingsMsg'
Learn More

Ryuvidine, Tocris Bioscience™

SETD8 inhibitor; also CDK4 inhibitor

Marke:  Tocris (Bio-Techne) 260910

Weitere Details : Netto-Gewicht : 0.00010kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411574

  • 252.13 € / 10 Milligramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Inhibitor of SETD8 protein lysine methyltransferase (PKMT) (IC 50 = 0.5 μM); suppresses H4K20 monomethylation in vitro. Also inhibits cyclin-dependent kinase (CDK) 4 (IC 50 = 6.0 μM at CDK4/cyclin D1). Induces S phase accumulation in HEK293T cells. Cytotoxic against a range of human cancer cells.
Spezifikation

Chemische Identifikatoren

265312-55-8
284.333
cdk4 inhibitor iii, ryuvidine, 2-methyl-5-4-methylphenyl amino benzothiazole-4,7-dione, 5-n-4-methylphenyl amino-2-methyl-4,7-dioxobenzothiazole, 2-methyl-5-p-tolylamino-benzothiazole-4,7-dione, 2-methyl-5-4-methylanilino-1,3-benzothiazole-4,7-dione, d07hpe, 2-methyl-5-4-methylphenyl amino-4,7-benzothiazoledione, 2-methyl-5-p-tolylamino benzo d thiazole-4,7-dione, 2-methyl-5-4-methylphenylamino-4,7-dihydrobenzothiazole-4,7-dione
2-methyl-5-(4-methylanilino)-1,3-benzothiazole-4,7-dione
C15H12N2O2S
HFPLHASLIOXVGS-UHFFFAOYSA-N
481747
CC1=CC=C(C=C1)NC2=CC(=O)C3=C(C2=O)N=C(S3)C

Spezifikation

Ryuvidine
265312-55-8
cdk4 inhibitor iii, ryuvidine, 2-methyl-5-4-methylphenyl amino benzothiazole-4,7-dione, 5-n-4-methylphenyl amino-2-methyl-4,7-dioxobenzothiazole, 2-methyl-5-p-tolylamino-benzothiazole-4,7-dione, 2-methyl-5-4-methylanilino-1,3-benzothiazole-4,7-dione, d07hpe, 2-methyl-5-4-methylphenyl amino-4,7-benzothiazoledione, 2-methyl-5-p-tolylamino benzo d thiazole-4,7-dione, 2-methyl-5-4-methylphenylamino-4,7-dihydrobenzothiazole-4,7-dione
CC1=CC=C(C=C1)NC2=CC(=O)C3=C(C2=O)N=C(S3)C
284.333
284.33
10mg
C15H12N2O2S
HFPLHASLIOXVGS-UHFFFAOYSA-N
2-methyl-5-(4-methylanilino)-1,3-benzothiazole-4,7-dione
481747
>98%
Videos