missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Benzyl-3-Isobutylpiperazin, 97+ %, Thermo Scientific™

Catalog Number p-7034604
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
444892-03-9
C15H24N2
232.371
MFCD03787915
HQMROSCVRBNRRZ-HNNXBMFYSA-N
s-1-benzyl-3-isobutylpiperazine, s-n4-benzyl-2-isobutylpiperazine, 3s-1-benzyl-3-2-methylpropyl piperazine, 3s-1-benzyl-3-isobutyl-piperazine, s-1-benzyl-3-isobutyl piperazine, 1-benzyl-3 s---isobutylpiperazine
17998949
(3S)-1-benzyl-3-(2-methylpropyl)piperazin
CC(C)CC1CN(CCN1)CC2=CC=CC=C2
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
444892-03-9
C15H24N2
232.371
MFCD03787915
HQMROSCVRBNRRZ-HNNXBMFYSA-N
s-1-benzyl-3-isobutylpiperazine, s-n4-benzyl-2-isobutylpiperazine, 3s-1-benzyl-3-2-methylpropyl piperazine, 3s-1-benzyl-3-isobutyl-piperazine, s-1-benzyl-3-isobutyl piperazine, 1-benzyl-3 s---isobutylpiperazine
17998949
(3S)-1-benzyl-3-(2-methylpropyl)piperazin
CC(C)CC1CN(CCN1)CC2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (S)-1-Benzyl-3-isobutylpiperazine
Menge 250 mg
Formelmasse 232.37
Reinheit (%) ≥97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'