missing translation for 'onlineSavingsMsg'
Learn More

S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium Tetrafluorborat, ≥ 98 %, Thermo Scientific™

Catalog Number p-7036155
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
255825-38-8
C10H16BF4N3OS
313.12
MFCD05664718
LHLFXDQURZVFLK-UHFFFAOYSA-N
tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, 2-dimethylamino dimethyliminio methyl sulfanyl pyridin-1-ium-1-olate tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate, s-2-pyridyl-n,n,n',n'-tetramethylthiuronium n-oxide tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate nmr, tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, dimethylamino-1-oxidopyridin-1-ium-2-yl sulfanyl-methylene-dimethyl-ammonium tetrafluoroborate
10543173
[Dimethylamino-(1-Oxidopyridin-1-ium-2-yl)Sulfanylmethyliden]-Dimethylazanium;Tetrafluorborat
F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
255825-38-8
C10H16BF4N3OS
313.12
MFCD05664718
LHLFXDQURZVFLK-UHFFFAOYSA-N
tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, 2-dimethylamino dimethyliminio methyl sulfanyl pyridin-1-ium-1-olate tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate, s-2-pyridyl-n,n,n',n'-tetramethylthiuronium n-oxide tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate nmr, tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, dimethylamino-1-oxidopyridin-1-ium-2-yl sulfanyl-methylene-dimethyl-ammonium tetrafluoroborate
10543173
[Dimethylamino-(1-Oxidopyridin-1-ium-2-yl)Sulfanylmethyliden]-Dimethylazanium;Tetrafluorborat
F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium tetrafluoroborate
Schmelzpunkt 111°C to 116°C
Menge 1 g
Empfindlichkeit Moisture sensitive
Formelmasse 314.13
Reinheit (%) ≥98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'