missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-(3-Fluorphenyl)propionsäure, 95%, Thermo Scientific™

Catalog Number p-7034405
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
723284-79-5
C9H10FNO2
183.182
MFCD04113689
GZNJUJNKZBHINS-QMMMGPOBSA-N
s-3-amino-3-3-fluorophenyl propanoic acid, s-3-amino-3-3-fluoro-phenyl-propionic acid, 3s-3-amino-3-3-fluorophenyl propanoic acid, h-beta-phe 3-f-oh, s-beta-3-fluoro-phenyl alanine, s-3-amino-3-3-fluorophenyl propionic acid, s-3-3-fluoro-phenyl-beta-alanine, h-d-phg 3-f-c*ch2 oh, 3-fluoro-l-beta-phenylalanine
738043
(3S)-3-Amino-3-(3-fluorphenyl)propansäure
C1=CC(=CC(=C1)F)C(CC(=O)O)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
723284-79-5
C9H10FNO2
183.182
MFCD04113689
GZNJUJNKZBHINS-QMMMGPOBSA-N
s-3-amino-3-3-fluorophenyl propanoic acid, s-3-amino-3-3-fluoro-phenyl-propionic acid, 3s-3-amino-3-3-fluorophenyl propanoic acid, h-beta-phe 3-f-oh, s-beta-3-fluoro-phenyl alanine, s-3-amino-3-3-fluorophenyl propionic acid, s-3-3-fluoro-phenyl-beta-alanine, h-d-phg 3-f-c*ch2 oh, 3-fluoro-l-beta-phenylalanine
738043
(3S)-3-Amino-3-(3-fluorphenyl)propansäure
C1=CC(=CC(=C1)F)C(CC(=O)O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (S)-3-Amino-3-(3-fluorophenyl)propionic acid
Formelmasse 183.18
Reinheit (%) 95%
Menge 250 mg
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'