missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-(4 -cyanophenyl)propionsäure, 95%, Thermo Scientific™

Catalog Number p-7034400
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
718596-77-1
C10H10N2O2
190.20
MFCD04117832
JFPLLJRLBHIJPS-UHFFFAOYNA-N
s-3-amino-3-4-cyanophenyl propanoic acid, s-3-amino-3-4-cyano-phenyl-propionic acid, 3s-3-amino-3-4-cyanophenyl propanoic acid, s-3-amino-3-4-cyanophenyl propionic acid, h-beta-phe 4-cn-oh, 4-cyano-l-beta-phenylalanine, s-beta-amino-4-cyanohydrocinnamic acid, s-3-amino-4-cyanophenyl propionic acid, s-3-amino-3-4-cyanophenyl propanoicacid, benzenepropanoic acid, b-amino-4-cyano-, bs
1491714
NC(CC(O)=O)C1=CC=C(C=C1)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
718596-77-1
C10H10N2O2
190.20
MFCD04117832
JFPLLJRLBHIJPS-UHFFFAOYNA-N
s-3-amino-3-4-cyanophenyl propanoic acid, s-3-amino-3-4-cyano-phenyl-propionic acid, 3s-3-amino-3-4-cyanophenyl propanoic acid, s-3-amino-3-4-cyanophenyl propionic acid, h-beta-phe 4-cn-oh, 4-cyano-l-beta-phenylalanine, s-beta-amino-4-cyanohydrocinnamic acid, s-3-amino-4-cyanophenyl propionic acid, s-3-amino-3-4-cyanophenyl propanoicacid, benzenepropanoic acid, b-amino-4-cyano-, bs
1491714
NC(CC(O)=O)C1=CC=C(C=C1)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (S)-3-Amino-3-(4-cyanophenyl)propionic acid
IUPAC-Name 3-amino-3-(4-cyanophenyl)propanoic acid
Formelmasse 190.2
Reinheit (%) 95%
Menge 250 mg
Empfindlichkeit Air Sensitive
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'