Learn More
(S)-ar-Curcumen, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | (S)-ar-Curcumene |
| CAS | 4176-06-1 |
| Summenformel | C15H22 |
| Menge | 50 mg |
| Synonym | (+)-Curcumene, (+)-ar-Curcumene, (+)-alpha-Curcumene, (S)-(+)-Curcumene, (S)-ar-Curcumene, (S)-alpha-Curcumene, S-Curcumene, ar-Curcumene, (+)-, d-ar-Curcumene, (+)alpha-Curcumene1-[(1S)-1,5-Dimethyl-4-hexen-1-yl]-4-methylbenzene, (+)-(S)-ar-Curcumene, 1-[(1S)-1,5-dimethyl-4-hexenyl]-4-methyl-benzene |
| InChI-Formel | InChI=1S/C15H22/c1-12(2)6-5-7-14(4)15-10-8-13(3)9-11-15/h6,8-11,14H,5,7H2,1-4H3/t14-/m0/s1 |
| SMILES | C[C@@H](CCC=C(C)C)c1ccc(C)cc1 |
| IUPAC-Name | 1-Methyl-4-[(2S)-6-methylhept-5-en-2-yl]benzol |
| Molekulargewicht (g/mol) | 202.34 |
| Formelmasse | 202.1722 |