missing translation for 'onlineSavingsMsg'
Learn More

S-Ethylthiopropionat, 97 %, Thermo Scientific™

Catalog Number p-7027865
Click to view available options
Menge:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2432-42-0
C5H10OS
118.194
MFCD00027016
HNEVHBHRLCAKKQ-UHFFFAOYSA-N
s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester
75513
S-Ethyl-propanthioat
CCC(=O)SCC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2432-42-0
C5H10OS
118.194
MFCD00027016
HNEVHBHRLCAKKQ-UHFFFAOYSA-N
s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester
75513
S-Ethyl-propanthioat
CCC(=O)SCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material S-Ethyl thiopropionate
Dichte 0.958
Siedepunkt 137°C to 138°C
Flammpunkt 27°C (80°F)
Geruch Stench
Brechungsindex 1.459
Menge 5 g
UN-Nummer UN1993
Beilstein 1740740
Formelmasse 118.2
Reinheit (%) 97%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'