missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(+)-2-Amino-1-Phenylpropyldiphenylphosphin, 97+%, Thermo Scientific Chemicals

Catalog Number p-7040540
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
341968-71-6
C21H22NP
319.39
MFCD11044864
JWZAIGGNEGTDMG-AURADERDNA-N
1s,2s-1-diphenylphosphino-1-phenylpropan-2-amine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphine, 1s,2s-2-amino-1-phenylpropyldiphenylphosphine, 1s,2s-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1s,2s-2-diphenylphosphino-1-methyl-2-phenylethylamine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphane, 1s,2s-1-diphenylphosphino-1-phenyl-2-propanamine, 1s,2s-2-amino-1-phenylpropyldiphenylphosphine,min, s,s-+-2-amino-1-phenylpropyldiphenylphosphine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphine, kanata purity
46176707
C[C@H](N)[C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
341968-71-6
C21H22NP
319.39
MFCD11044864
JWZAIGGNEGTDMG-AURADERDNA-N
1s,2s-1-diphenylphosphino-1-phenylpropan-2-amine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphine, 1s,2s-2-amino-1-phenylpropyldiphenylphosphine, 1s,2s-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1s,2s-2-diphenylphosphino-1-methyl-2-phenylethylamine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphane, 1s,2s-1-diphenylphosphino-1-phenyl-2-propanamine, 1s,2s-2-amino-1-phenylpropyldiphenylphosphine,min, s,s-+-2-amino-1-phenylpropyldiphenylphosphine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphine, kanata purity
46176707
C[C@H](N)[C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (S,S)-(+)-2-Amino-1-phenylpropyldiphenylphosphine
Schmelzpunkt 100°C to 107°C
Menge 250 mg
Empfindlichkeit Air Sensitive
Optische Drehung +140° (c=0.5 in Chloroform)
IUPAC-Name (2-Amino-1-phenylpropyl)diphenylphosphan
Formelmasse 319.38
Reinheit (%) ≥97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'