Learn More
Invitrogen™ Human S100A10, Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP75716
Weitere Details : CAS : 69-74-9 Netto-Gewicht : 0.01000kg
Beschreibung
S100A10 (human) Recombinant Protein, RP-75716, from Invitrogen.
This product contains purified recombinant human S100A10 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.Spezifikation
P60903 | |
Control, Western Blot | |
6281 | |
100μg | |
N/A | |
RUO | |
S100A10 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
0.10 mg/mL | |
PBS with 50% glycerol and no preservative | |
Human S100A10 | |
E. Coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human S100A10 | |
Human | |
Hu | |
Recombinant | |
Recombinant |