missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550783
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total S100A14 protein.
S100A14 Polyclonal antibody specifically detects S100A14 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
S100A14 | |
Polyclonal | |
Unconjugated | |
S100a14 | |
1110013O05Rik; BCMP84; breast cancer membrane protein 84; Gm1020; Protein S100-A14; S100 calcium binding protein A14; S100 calcium binding protein A15; S100 calcium-binding protein A14; S100A14; S100a15; S114 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57402 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9HCY8 | |
S100a14 | |
Full length fusion protein of Human S100A14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |