missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564022
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EMKELLHKEL PSFVGEKVDE EGLKKLMGSL DENSDQQVD Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 56%.
S100A2 Polyclonal antibody specifically detects S100A2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
S100A2 | |
Polyclonal | |
Unconjugated | |
S100A2 | |
CAN19; EG628324; predicted gene, EG628324; Protein S100-A2; Protein S-100L; S100 calcium binding protein A2; S100 calcium-binding protein A2; S100a2; S100L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P29034 | |
S100A2 | |
Recombinant protein corresponding to Human S100A2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only