missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550099
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total S100A5 protein.
S100A5 Polyclonal antibody specifically detects S100A5 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
S100A5 | |
Polyclonal | |
Unconjugated | |
S100A5 | |
Protein S100-A5; Protein S-100D; S100 calcium binding protein A5; S100 calcium-binding protein A4; S100 calcium-binding protein A5; S100a5; S100D; S100D9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20199, 295211, 6276 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P33763, P63083, P63084 | |
S100A5 | |
Synthesized peptide derived from internal of human S100A5. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |