missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A7A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA568288
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
S100A7A Polyclonal antibody specifically detects S100A7A in Human samples. It is validated for Western Blot.Spezifikation
S100A7A | |
Polyclonal | |
Unconjugated | |
S100A7A | |
NICE-2; Protein S100-A7A; S100 calcium binding protein A15; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100A7A; S100A7f; S100A7L1 | |
Rabbit | |
affinity chromatography | |
RUO | |
338324 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q86SG5 | |
S100A7A | |
Synthesized peptide derived from human S100A7A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |