missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A7L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563496
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SDMDYLSNAL EKKDDNKDKK VNYSE Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 41%.
S100A7L2 Polyclonal antibody specifically detects S100A7L2 in Human samples. It is validated for Immunohistochemistry.Spezifikation
S100A7L2 | |
Polyclonal | |
Unconjugated | |
S100A7L2 | |
protein S100-A7-like 2; S100 calcium binding protein A7 like 2; S100 calcium binding protein A7-like 2; S100 calcium-binding protein A7-like 2; S100a7b; S100A7L2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
645922 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5SY68 | |
S100A7L2 | |
Recombinant protein corresponding to Human S100A7L2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only