Learn More
S100PBP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555585
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PDLFKLPQLS TSSGHGPAHT KPLNRRSVLE KNLIKVTVAP FNPTVCDALL DKDETDSSKD TEKLSSLGEE MREDGLSPNE SKLCTESEGI SPNNS Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 72%.
S100PBP Polyclonal antibody specifically detects S100PBP in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
S100PBP | |
Polyclonal | |
Unconjugated | |
S100PBP | |
4930429A08Rik; AI851343; hypothetical protein LOC539983; RGD1564943; S100P binding protein; S100P binding protein 1; S100P-binding protein; S100P-binding protein Riken; S100pbp; S100PBPR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64766 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BU1 | |
S100PBP | |
Recombinant protein corresponding to Human S100PBP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only