Learn More
SLC13A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561773
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HNENLNGVPS ITNPIKTANQ HQGKKQHPSQ EKPQVLTPSP RKQKLNRKYR SHHDQMICKC LSLSI Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.
SLC13A4 Polyclonal antibody specifically detects SLC13A4 in Human samples. It is validated for Immunohistochemistry.Spezifikation
SLC13A4 | |
Polyclonal | |
Unconjugated | |
Slc13a4 | |
9630060C05Rik; Na(+)/sulfate cotransporter SUT-1; NAS2; SLC13A4; sodium sulfate cotransporter-2; solute carrier family 13 (sodium/sulfate symporter), member 4; solute carrier family 13 (sodium/sulfate symporters), member 4; solute carrier family 13 (sodium/sulphate symporters), member 4; solute carrier family 13 member 4; sulphate transporter 1; SUT1; SUT-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26266 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKG4 | |
Slc13a4 | |
Recombinant protein corresponding to Human S13A4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only