missing translation for 'onlineSavingsMsg'
Learn More

S7A6O Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA559614

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15717471

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: AEALVLACKR LRSDAVESAA QKTSEGLERA AENNVFHLVA TVCSQEEPVQ PLLREVLRPS RDSQQRVRRN LRASAREVRQ EGRYRVLS Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

S7A6O Polyclonal antibody specifically detects S7A6O in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

S7A6O
Polyclonal
Unconjugated
SLC7A6OS
ADAMS proteinase-related protein; Probable RNA polymerase II nuclear localization protein SLC7A6OS; protein SLC7A6OS; SLC7A6OS; solute carrier family 7 member 6 opposite strand; solute carrier family 7 member 6 opposite strand transcript; solute carrier family 7, member 6 opposite strand
Rabbit
Antigen affinity chromatography
RUO
84138
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96CW6
SLC7A6OS
Recombinant protein corresponding to Human S7A6O
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only