Learn More
Asecos™ SICHERHEITSSCHRANK TYP 90 MIT INNENEINR G
Marke: Asecos™ 30231-001-30269
Weitere Details : Netto-Gewicht : 261.00000kg
Beschreibung
Type 90 safety storage cabinet S-Pegasus-90, S90.196.060.WDAC including six pull-out drawers, plastic laminated sheet steel RAL 7035.- Convenient handling - one-hand operation for double-door cabinets (AGT)
- Always closed - serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds. Acoustic and visual signal before closing
- Safety elements assembled outside the storage compartment for increased protection against corrosion, easy to clean
- Door open arrest system (standard), convenient and safe removal or storage of containers
- Doors lockable with cylinder locking (integration in a locking system possible)
- Adjusting aids to compensate for uneven floor
- Integrated air ducts ready for connection (NW 75) to a technical exhaust system
- Fire resistance 90 minutes
- GS approval, CE conformity
- EN 14727 conformity (laboratory furniture)
Technical data model S90.196.060.WDACDimensions (w x d x h), exterior: 600mm x 615mm x 1968mmDimensions (w x d x h), interior: 450mm x 520mm x 1740mmHeight including transport base: 2090mmDepth with open doors : 1136mmWeight (kg): 255Extraction air (NW): 75Air exchange 10 times (m3/h): 4.1Pressure drop inside the cabinet (Pa): <5δρ total (EK5/AK4 09-10) (Pa): <5Maximum load (kg): 600Distributed load (max. N/mm2): 0.228