missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SAK 3

Description
SAK 3 is a potent CaV3.1 and CaV3.3 activator. Enhances CaV3.1 and CaV3.3, but not CaV3.2 activity at concentrations of 0.01 - 10 M in neuro2A cells in vitro. Elevates ACh levels in hippocampus. Rescues impaired memory-related behaviors, promotes adult neurogenesis and displays antidepressant-like effects in OBX mice. Orally bioavailable.
Specifications
Specifications
| Chemischer Name oder Material | Ethyl 8′-methyl-2′,4-dioxo-2-(piperidin-1-yl)-2′H-spiro[cyclopent[2]ene-1,3′-imidazo[1,2-a]pyridine]-3-carboxylate |
| CAS | 1256269-87-0 |
| Menge | 25 mg |
| Ziel | Cav3.x Channel Activators |
| Summenformel | C20H23N3O4 |
| Reinheit | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?