Learn More
Thermo Scientific™ Sample Cuvettes for Eutech™ TN-100 Turbidity Meter
Product Code. 17425499
Change view
Click to view available options
For Use With (Equipment):
Eutech TN-100 Turbidity Meter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17425499 Eutech TN-100 Turbidity Meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17425499 Supplier Thermo Scientific™ Supplier No. 01X638601
  • ONLINE EXCLUSIVE PRICE
    133.00 €
    121.00 € / 1 Stück
    Sparen 12.00 €
    9% Rabatt
 Restricted Item
Verfügbar ab: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
TN-100 meter is an easy-to-use turbidimeter that meets the rigor of rugged field applications.
Sample Cuvettes for Eutech™ TN-100 Turbidity Meter are borosilicate glass vials with light-shield screwcaps, fill line and indexing marks.The cuvettes come in pack of four.
Features:
  • Pack of 4 sample cuvettes for TN-100 Turbidity Meter
TRUSTED_SUSTAINABILITY
Specifications
Type Cuvette
For Use With (Equipment) Eutech TN-100 Turbidity Meter
Description Sample Cuvettes
Quantity 4 Pack