Learn More
IBA Lifesciences SARS-CoV-2 Peptide KIADYNYKL (HLA-A*0201)
Alle ansehen IBA Lifesciences Produkte
Description
- SARS-CoV-2 peptide KIADYNYKL (HLA-A*0201) is a single peptide for stimulation of T cells.
- The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
Specifications
| Reinheit | >85% |
| Endotoxin-Gehalt | Free |
| Format | Lyophilisiertes Pulver |
| Menge | 1 mg |
| Forschungskategorie | T cell stimulation |
Nur für Forschungszwecke.